Merck
  • Elevated expression of cyclooxygenase (COX)-2 in oral squamous cell carcinoma--evidence for COX-2 induction by areca quid ingredients in oral keratinocytes.
Elevated expression of cyclooxygenase (COX)-2 in oral squamous cell carcinoma--evidence for COX-2 induction by areca quid ingredients in oral keratinocytes.
Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology (2003-09-13)
Deh-Wei Tang, Shu-Chun Lin, Kuo-Wei Chang, Chin-Wen Chi, Che-Shoa Chang, Tsung-Yun Liu
ABSTRACT
Elevated expression of cyclooxygenase (COX)-2 has been demonstrated in several human cancers. Whether COX-2 is up-regulated in areca quid (AQ) related oral squamous cell carcinoma (OSCC) is unknown and the potential of AQ ingredients to induce COX-2 expression has not been studied. COX-2 expression was analyzed by immunohistochemistry and RT-PCR in oral tissues. The COX-2 mRNA and protein induction potential of AQ ingredients were analyzed by real-time RT-PCR and Western blotting in normal human oral keratinocyte (NHOK). COX-2 protein expression was significantly higher (P < 0.01) in OSCC (n = 27) as compared to their adjacent non-cancerous matched tissue (NCMT). COX-2 protein was nearly undetectable in control normal oral mucosa. The level of COX-2 mRNA was markedly elevated in 63% (12/19) of OSCC compared to NCMT. Hydroxychavicol induced COX-2 mRNA and protein expression in NHOK. COX-2 protein as well as mRNA expression were significantly enhanced in OSCC as compared to NCMT. Hydroxychavicol, a unique ingredient in AQ, induced COX-2 expression in NHOK, which highlighted early involvement of COX-2 in AQ-associated oral oncogenesis.