Merck
  • [Comprehensive study of lead pollution in atmospheric aerosol of Shanghai].
[Comprehensive study of lead pollution in atmospheric aerosol of Shanghai].
Huan jing ke xue= Huanjing kexue (2006-07-21)
Gui-lin Zhang, Ming-guang Tan, Xiao-lin Li, Yuan-xun Zhang, Wei-sheng Yue, Jian-min Chen, Yin-song Wang, Ai-guo Li, Yan Li, Yuan-mao Zhang, Zhu-ci Shan
ABSTRACT
The lead contamination, lead species and source assignment were studied by a combination of several analytical techniques such as Proton-induced X-ray emission analysis (PIXE), Proton microprobe (micro-PIXE), Inductively coupled plasma-mass spectrometry (ICP-MS) and extended X-ray absorption fine structure (EXAFS) techniques. The results indicate that the lead concentration in the air of Shanghai gradually decreased over the last years. The atmospheric lead concentration of PM10 in the winter of 2002 was 369 ng x m(-3), which had declined by 28% in 2001, and in the winter of 2003 it decreased further to 237 ng x m(-3). The main lead species in the samples collected in the winter of 2003 were probably PbCl2, PbSO4 and PbO. The source apportionment was calculated in terms of the combination of lead isotope ratios and lead mass balance method, assisted by single particle analysis with micro-PIXE and pattern recognition. The results suggest that the major contributors of atmospheric lead pollution in Shanghai are the coal combustion dust; the metallurgic dust and vehicle exhaust particles, with a contribution around 50%, 35% and 15%, respectively. It probably is the first time to give a city a quantitative estimation of lead pollution contribution from emission sources. The influence from leaded gasoline was still present in the atmosphere by four or five years after the phasing out of leaded gasoline.
Sigma-Aldrich
Lead(II) sulfate, 99.99% trace metals basis
Sigma-Aldrich
Lead(II) sulfate, 98%