Merck
  • Enantioselective synthesis of 3,3-disubstituted piperidine derivatives by enolate dialkylation of phenylglycinol-derived oxazolopiperidone lactams.
Enantioselective synthesis of 3,3-disubstituted piperidine derivatives by enolate dialkylation of phenylglycinol-derived oxazolopiperidone lactams.
The Journal of organic chemistry (2007-05-10)
Mercedes Amat, Oscar Lozano, Carmen Escolano, Elies Molins, Joan Bosch
ABSTRACT
The stereochemical outcome of the enolate dialkylation of simple phenylglycinol-derived oxazolopiperidone lactams is studied. High stereoselectivities in the generation of the quaternary stereocenter are obtained by the appropriate choice of both the configuration of the starting lactam and the order of introduction of the substituents. The usefulness of the methodology is illustrated by the conversion of some of the dialkylated lactams into known synthetic precursors of alkaloids and by the total synthesis of (-)-quebrachamine.
Sigma-Aldrich
N-(2-Hydroxyethyl)aniline, 98%