Merck
  • Structural insight into the quinolone-DNA cleavage complex of type IIA topoisomerases.
Structural insight into the quinolone-DNA cleavage complex of type IIA topoisomerases.
Nature structural & molecular biology (2009-05-19)
Ivan Laponogov, Maninder K Sohi, Dennis A Veselkov, Xiao-Su Pan, Ritica Sawhney, Andrew W Thompson, Katherine E McAuley, L Mark Fisher, Mark R Sanderson
ABSTRACT
Type II topoisomerases alter DNA topology by forming a covalent DNA-cleavage complex that allows DNA transport through a double-stranded DNA break. We present the structures of cleavage complexes formed by the Streptococcus pneumoniae ParC breakage-reunion and ParE TOPRIM domains of topoisomerase IV stabilized by moxifloxacin and clinafloxacin, two antipneumococcal fluoroquinolones. These structures reveal two drug molecules intercalated at the highly bent DNA gate and help explain antibacterial quinolone action and resistance.