Merck
  • Determination of ipriflavone in human plasma by LC-MS and its application in a pharmacokinetic study.
Determination of ipriflavone in human plasma by LC-MS and its application in a pharmacokinetic study.
Biomedical chromatography : BMC (2011-05-20)
Changhong Yun, Li Ding, Ye Leng, He Zhu, Aidong Wen, Lin Yang
ABSTRACT
A sensitive liquid chromatography-mass spectrometric method was developed for the quantification of ipriflavone in human plasma. The method utilized liquid-liquid extraction of plasma with ethyl acetate. A gradient elution was performed on a Hedera ODS-2 column (150×2.1 mm i.d., 5 µm), using a mobile phase consisting of 0.1% formic acid solution and methanol at a flow rate of 0.5 mL/min. The single quadrupole mass spectrometer was operated in selected-ion monitoring mode via positive electrospray ionization interface detecting m/z 239.1 and 285.1 for ipriflavone and diazepam (the internal standard), respectively. To improve the selectivity and sensitivity, the fragment ion m/z 239.1, which was produced by in-source collision-induced dissociation, was chosen as the quantitative ion for ipriflavone. The method was fully validated and applied to a pharmacokinetic study of ipriflavone. After oral administration of a single 200 mg ipriflavone tablet, the C(max,) AUC(0-72 h) , t(1/2) and T(max) were 6.3±6.3 ng/mL, 80.0±69.1 µg h/L, 23.0±8.6 h and 3.4±2.1 h, respectively.
Sigma-Aldrich
7-Isopropoxy-3-phenyl-4H-1-benzopyran-4-one, 97%