Merck
  • [Structural and functional features of the rats' prefrontal cortex following 14-day suspension].
[Structural and functional features of the rats' prefrontal cortex following 14-day suspension].
Aviakosmicheskaia i ekologicheskaia meditsina = Aerospace and environmental medicine (2011-09-16)
G A Romanova, A A Pal'tsyn, F M Shakova, N B Konstantinova, M V Baranov, V M Baranov
ABSTRACT
Effects of 14-d suspension on the processes of rats' nervous cells conjugation in the brain cognitive cortex were studied. Signs of cognitive functions degradation and decrease of the number of conjugated nervous cells were found in the experimental group after suspension. This evidence are qualified as a negative effect of suspension on memory and physiological regeneration of neurons.
Sigma-Aldrich
Cresyl Violet acetate, certified by the BSC
Sigma-Aldrich
Cresyl Violet perchlorate