Merck
  • Selenium containing imidazolium salt in designing single source precursors for silver bromide and selenide nano-particles.
Selenium containing imidazolium salt in designing single source precursors for silver bromide and selenide nano-particles.
Dalton transactions (Cambridge, England : 2003) (2013-01-08)
Hemant Joshi, Kamal Nayan Sharma, Ved Vati Singh, Pradhumn Singh, Ajai Kumar Singh
ABSTRACT
The AgBr and Ag(2)Se nanoparticles (NPs) have been synthesized for the first time from two single source precursors ([Ag(2)(L)(2)Br(2)] (1) and [Ag(L-HBr)(2)]BF(4) (2) respectively) designed using the same ligand 3-benzyl-1-(2-phenylselanyl-ethyl)-3H-imidazolium bromide (L). The ODE-ODA-OA (1 : 1 : 2) and TOP-OA (1 : 2) are most suitable solvents for thermolysis of 1 and 2 respectively, resulting in the NPs. The composition of the solvent used in thermolysis affects the purity of NPs. The bonding of L in 1 is unique, as it has a pre-carbene site intact.
Sigma-Aldrich
Silver bromide, 99%