Merck
  • Giant and broad-band absorption enhancement in colloidal quantum dot monolayers through dipolar coupling.
Giant and broad-band absorption enhancement in colloidal quantum dot monolayers through dipolar coupling.
ACS nano (2013-01-10)
Pieter Geiregat, Yolanda Justo, Sofie Abe, Stijn Flamee, Zeger Hens
ABSTRACT
The absorption cross section of colloidal quantum dots in close-packed monolayers shows a 4 (CdSe) to 5-fold (PbS) enhancement compared to quantum dots in a dilute dispersion. Quantitative agreement is demonstrated between the value and the size dependence of the enhancement and theoretical model predictions based on dipolar coupling between neighboring quantum dots. This collective optical behavior offers a new degree of freedom in the custom design of optical properties for electro-optical devices.
Sigma-Aldrich
Lead(II) sulfide, 99.9% trace metals basis