Merck
[Chemical constituents from Coptis chinensis].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2013-03-05)
Zhi-Feng Li, Qi Wang, Yu-lin Feng, Xiao-jian Luo, Mei-mei Fan, Shi-lin Yang
ABSTRACT
To study the chemical constituents of Coptis chinensis. The compounds were separated and purified by column chromatography and their structures were identified by spectroscopic methods. Seven compounds were isolated from the chloroform extract of Coptis chinensis, whose structures were determined as worenine (1),8,9-dihydroxy-1,5,6, 10b-tetrahydro-2H-pyrrolo[2,1 -alpha] isoquinolin-3-one (2),(+/-) -5,5'-dimethoxy-lariciresinol (3),3,4-dihydroxy-phenylethylalcohol(4), methyl-5-O-feruloylquinate (5), ethyl-5-O- feruloylquinate (6), apocynol (7). Compounds (3) is isolated from this genus for the first time, compounds (5), (6), (7) are isolated from this plant for the first time.
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Guaiacol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Guaiacol, oxidation indicator
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG