Merck
  • The effect of sodium bicarbonate upon urinary citrate excretion in calcium stone formers.
The effect of sodium bicarbonate upon urinary citrate excretion in calcium stone formers.
Urology (2013-04-23)
Vivian Barbosa Pinheiro, Alessandra Calábria Baxmann, Hans-Göran Tiselius, Ita Pfeferman Heilberg
ABSTRACT
To evaluate the effects of oral sodium bicarbonate (NaBic) supplementation upon urinary citrate excretion in calcium stone formers (CSFs). Sixteen adult calcium stone formers with hypocitraturia were enrolled in a randomized, double-blind, crossover protocol using 60 mEq/day of NaBic during 3 days compared to the same period and doses of potassium citrate (KCit) supplementation. Blood and 24-hour urine samples were collected at baseline and during the third day of each alkali salt. NaBic, similarly to KCit supplementation, led to an equivalent and significant increase in urinary citrate and pH. Compared to baseline, NaBic led to a significant increase in sodium excretion without concomitant increases in urinary calcium excretion, whereas KCit induced a significant increase in potassium excretion coupled with a significant reduction in urinary calcium. Although NaBic and KCit both reduced calcium oxalate supersaturation (CaOxSS) significantly vs baseline, KCit reduced calcium oxalate supersaturation significantly further vs NaBic. Both KCit and NaBic significantly reduced urinary phosphate and increased calcium phosphate supersaturation (CaPSS) compared to baseline. Finally, a significantly higher sodium urate supersaturation (NaUrSS) was observed after the use of the 2 drugs. This short-term study suggests that NaBic represents an effective alternative for the treatment of hypocitraturic calcium oxalate stone formers who cannot tolerate or afford the cost of KCit. In view of the increased sodium urate supersaturation, patients with pure uric acid stones and high urate excretion may be less suited for treatment with NaBic.
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Potassium citrate tribasic monohydrate, meets USP testing specifications
Sigma-Aldrich
Potassium citrate tribasic monohydrate, ≥98% (GC/titration)
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Uric acid, ≥99%, crystalline
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Potassium citrate tribasic monohydrate, suitable for cell culture
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Potassium citrate tribasic monohydrate, purum p.a., ≥99.0% (NT)
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium citrate tribasic monohydrate, meets analytical specification of Ph. Eur., BP, FCC, E332, 99-100.5% (calc with ref. to anhyd. subst.)
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Supelco
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Calcium oxalate monohydrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder