Merck
A mechanistic overview of TET-mediated 5-methylcytosine oxidation.
Biochemical and biophysical research communications (2013-06-04)
V K Chaithanya Ponnaluri, Jaroslaw P Maciejewski, Mridul Mukherji
ABSTRACT
Methylation of DNA at the carbon-5 position of cytosine plays crucial roles in the epigenetic transcriptional silencing during metazoan development. Recent identification of Ten-Eleven Translocation (TET)-family demethylases have added a new dimension to dynamic regulation of 5-methylcytosine (5mC), and thus, inheritable and somatic gene silencing. The interest in hematology was particularly stimulated by the recent discovery of TET2 mutations in myeloid malignancies which were proven to be leukemogenic in murine knockout models. The TET-family enzymes are Fe(II), 2-oxoglutarate-dependent oxygenases and catalyze demethylation of 5mC by iterative oxidation reactions. In the last decade results from numerous studies have established a key role for these enzymes in epigenetic transcriptional regulation in eukaryotes primarily by hydroxylation reactions. The TET catalyzed hydroxylation and dehydration reactions in the mammalian system exemplify the diversity of oxidation reactions catalyzed by Fe(II), 2-oxoglutarate-dependent oxygenases, and suggest an existence of other types of oxidation reactions catalyzed by these enzymes in the eukaryotes, which are so far only documented in prokaryotes. Here, we review the TET-mediated 5mC oxidation in light of the putative reaction mechanism of Fe(II), 2-oxoglutarate-dependent oxygenases.
Sigma-Aldrich
5-Methylcytosine hydrochloride, ≥99%
Sigma-Aldrich
α-Ketoglutaric acid potassium salt, ≥98% (enzymatic)