Merck
[New dyes containing gallocyanine for staining histological sections].
Arkhiv patologii (1986-01-01)
L A Faustov, A I Edemskiĭ
ABSTRACT
The methods of histological slides staining with dyes containing gallocyanin are proposed.
Sigma-Aldrich
Gallocyanine, Dye content 90 %