- Hydrophobic labelling of spectrin in erythrocytes using arylisothiocyanates.
Hydrophobic labelling of spectrin in erythrocytes using arylisothiocyanates.
Biomedica biochimica acta (1987-01-01)
A F Sikorski, M Kuczek, Z Nyczka, Z J Kubiak
PMID3579888
ABSTRACT
Labelling of spectrin with the hydrophobic probes 2-naphthylisothiocyanate and 4-[125I]-iodophenylisothiocyanate dispersed in lipid vesicles was studied. Although spectrin is a peripheral protein of the erythrocyte membrane, it was found to be labelled as a result of incubation of washed red blood cells with hydrophobic arylisothiocyanates.