Merck
  • Improved detection of oculomycoses using induced fluorescence with cellufluor.
Improved detection of oculomycoses using induced fluorescence with cellufluor.
Ophthalmology (1986-03-01)
J E Sutphin, N M Robinson, K R Wilhelmus, M S Osato
ABSTRACT
Cellufluor, a fluorochrome with affinity for chitin and cellulose, was retrospectively compared to gram and giemsa stains for detection of human oculomycoses. From culture-proven specimens, this chemofluorescent stain detected hyphae or yeasts on 50% of smears previously considered negative. Cellufluor is a useful adjunct in the detection and confirmation of fungal elements from ocular specimens.