Merck
  • A benzodiazepine receptor antagonist decreases sleep and reverses the hypnotic actions of flurazepam.
A benzodiazepine receptor antagonist decreases sleep and reverses the hypnotic actions of flurazepam.
Science (New York, N.Y.) (1983-01-28)
W B Mendelson, M Cain, J M Cook, S M Paul, P Skolnick
ABSTRACT
The benzodiazepine receptor antagonist 3-hydroxymethyl-beta-carboline, which blocks several of the pharmacological actions of benzodiazepines, induces a dose-dependent increase in sleep latency in the rat. Furthermore, at a low dose that by itself does not affect sleep, 3-hydroxymethyl-beta-carboline blocks sleep induction by a large dose of flurazepam. The benzodiazepine receptor may play a role in both the physiological regulation and pharmacological induction of sleep.