Merck
  • Effect of ergovaline, loline, and dopamine antagonists on rat pituitary cell prolactin release in vitro.
Effect of ergovaline, loline, and dopamine antagonists on rat pituitary cell prolactin release in vitro.
American journal of veterinary research (1994-05-01)
J R Strickland, D L Cross, G P Birrenkott, L W Grimes
ABSTRACT
Cultured rat pituitary cells were studied to: determine the effects of ergovaline and loline on in vitro prolactin release; delineate the agonistic activity of these alkaloids at the D2 dopamine receptor, using 2 selective D2 dopamine receptor antagonists; and compare the efficacy of 2 dopamine receptor antagonists in reversing effects of the treatments on in vitro prolactin secretion. Ergovaline reduced in vitro prolactin release by at least 40% (P < 0.05) at concentrations of 10(-4), 10(-6), and 10(-8) M. However, loline reduced (P < 0.05) prolactin release only at the highest concentration, 10(-4) M. Two standard dopamine agonists, dopamine and alpha-ergocryptine, were used to verify that the inhibitory control mechanisms of in vitro prolactin release were intact. Both compounds reduced prolactin release by at least 40% for concentrations of 10(-4), 10(-6), or 10(-8) M. Selective D2 dopamine receptor antagonists (10(-6) M), domperidone and sulpiride, reversed (P < 0.05) the effect of loline on in vitro prolactin release. However, only domperidone (10(-6) M) was able to reverse (P < 0.05) the effect of ergovaline and only at the lowest ergovaline concentration (10(-8) M). Domperidone was more effective (P < 0.05) in reversing the prolactin-suppressing effect of alpha-ergocryptine than was sulpiride. The dose-response curve for domperidone (cubic fit, P < 0.0001) indicated a threshold concentration (10(-7) M) for reversal of alpha-ergocryptine's (10(-8) M) effect on prolactin release. However, at similar concentration of sulpiride (quadratic fit, P < 0.007), a threshold level was not obtained.(ABSTRACT TRUNCATED AT 250 WORDS)