Merck
  • [Salicylism and glaucoma: reciprocal augmentation of the toxicity of acetazolamide and acetylsalicylic acid].
[Salicylism and glaucoma: reciprocal augmentation of the toxicity of acetazolamide and acetylsalicylic acid].
Journal francais d'ophtalmologie (1999-04-30)
E Hazouard, M Grimbert, A P Jonville-Berra, M C De Toffol, A Legras
ABSTRACT
We report a case of salicylism caused by association of acetylsalicylic acid (ASA) and acetazolamide. A 50-year-old woman without chronic renal insufficiency, was admitted for confusion, cerebellar ataxia associated with hyperchloremic acidosis. She was treated with acetazolamide for simple glaucoma and since 14 days with ASA for acute pericarditis. Clinical features evoked salicylism despite low serum salicylate level: 178 mg/l. The carbonic anhydrase inhibitor explained the biochemical pattern. Acetazolamide-induced acidosis enhances the non ionized form of salicylate which crosses the biological membrane more rapidly and better than the ionized form. The tissular tropism of non ionized salicylate increases and cerebral toxicity may occur with low serum level of ASA. ASA also enhances the free form of acetazolamide.
Product Description
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Salicylic acid, ≥99%, FG