Merck
  • Gas-chromatographic and mass-spectrometric detection of low-molecular-weight aliphatic alcohols in urine of normal individuals and patients with diabetes mellitus.
Gas-chromatographic and mass-spectrometric detection of low-molecular-weight aliphatic alcohols in urine of normal individuals and patients with diabetes mellitus.
Clinical chemistry (1975-08-01)
H M Liebich, O Al-Babbili, A Zlatkis, K Kim
ABSTRACT
We studied the aliphatic alcohols in 100 urines from 25 patients with diabetes mellitus under treatment with insulin, oral antidiabetic medication, or special diet. The procedure involves adsorption of the low-molecular-weight urinary metabolites on a porous polymer of 2,6-diphenyl-p-phenylene oxide (Tenax GC), gas-chromatographic separation, mass spectrometric identification, and mass fragmentographic representation of the primary alcohols by a computer. The concentrations of ethanol, n-propanol, isobutanol, n-butanol, and isopentanol are increased as compared with urine from normal persons.
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Sigma-Aldrich
1-Propanol, suitable for HPLC, ≥99.9%
Supelco
1-Propanol, analytical standard
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
1-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
1-Propanol, ≥99% (GC), purum