Merck
Vitamin A and zinc status in patients on maintenance haemodialysis.
Nephrology (Carlton, Vic.) (2005-10-14)
Poliana Coelho Cabral, Alcides da Silva Diniz, Ilma Kruze Grande de Arruda
ABSTRACT
The objective of this study was to assess the vitamin A and zinc serum levels in patients undergoing haemodialysis (HD) in the city of Recife, in the north-eastern region of Brazil. The study comprised 55 patients and 28 healthy controls. The retinol and zinc serum concentrations were analysed by using high-performance liquid chromatography (HPLC) and atomic absorption spectrophotometry, respectively. The mean retinol serum concentration in patients (2.50 +/- 0.86 micromol/L) was significantly greater (P < 0.001) than that found in controls (1.26 +/- 0.86 micromol/L). The retinol serum levels in the patients were as follows: 47.3% of the patients had elevated levels (>/= 2.24 and < 3.50 micromol/L); 16.4% of the patients had serum levels >/= 3.50 micromol/L, which indicated hypervitaminosis; and 9.1% of the patients had serum levels below the normal range (<1.05 micromol/L), a rate that among the controls was 42.9% (P < 0.01). In regard to zinc, the serum levels found in the patients (10.59 +/- 3.12 micromol/L) were similar to those found in the controls (11.43 +/- 2.82 micromol/L) (P > 0.05). Although 49.1% of the patients and 35.7% of the controls were classified as deficient in zinc, this difference was not statistically significant (P > 0.05). The results identified a high prevalence of zinc deficiency in the groups studied, and point to a trend towards more elevated retinol serum levels in patients undergoing dialysis as compared with those in healthy controls.
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
Sigma-Aldrich
DL-Homocysteine, ≥95% (titration)
Sigma-Aldrich
Zinc, powder, <150 μm, 99.995% trace metals basis
Sigma-Aldrich
Zinc, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Zinc, purum, powder
Sigma-Aldrich
Zinc, granular, 30-100 mesh, 99%
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Zinc, mossy, ≥99%
Sigma-Aldrich
trans-4-Hydroxy-L-proline, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn2+ in THF, highly reactive Rieke®metal
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg
Sigma-Aldrich
Retinol, ≥95.0% (HPLC), ~2700 U/mg
Supelco
4-Methylphenol solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
SAFC
L-Proline
Supelco
trans-4-Hydroxy-L-proline, analytical standard
Supelco
L-Proline, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
Glycolic acid solution, technical grade, 70 wt. % in H2O
Sigma-Aldrich
Glycolic acid, ReagentPlus®, 99%
Sigma-Aldrich
Glycolic acid, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Glycolic acid solution, high purity, 70 wt. % in H2O
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%