Merck
  • Longitudinal hair chromium profiles of elderly subjects with normal glucose tolerance and type 2 diabetes mellitus.
Longitudinal hair chromium profiles of elderly subjects with normal glucose tolerance and type 2 diabetes mellitus.
Metabolism: clinical and experimental (2006-12-13)
Janez Stupar, Matjaz Vrtovec, Franci Dolinsek
ABSTRACT
Longitudinal hair chromium (H-Cr) profiles in a group of patients with type 2 diabetes mellitus (n = 59; age, 62 +/- 9 years) and healthy elderly (control) subjects (n = 49; age, 59 +/- 10 years) matched by age and sex were measured by solid sampling electrothermal atomic absorption spectrometry, providing data on the magnitude of variation of Cr content along the hair length. H-Cr average (H-Cr(av)) and H-Cr proximal (H-Cr(pr))(.), relating to the average Cr content of the whole hair and the proximal 3-mm hair length, respectively, were also obtained. No significant difference between the healthy and diabetic group was found in mean H-Cr(av) or H-Cr(pr) contents (248 +/- 108 vs 247 +/- 143 and 233 +/- 98 vs 278 +/- 195 ng/g, respectively. However, women in the control group had significantly lower H-Cr values (P < .01) compared with men, but this difference was absent in the diabetic population. The distribution of log H-Cr(pr) values in the control population displayed a Gaussian shape, in contrast to the substantially wider distribution, skewed toward lower H-Cr(pr) values, observed in the diabetic group. The magnitude of variation in H-Cr content in the patient group over an interval of approximately 2 to 3 months (time of growth of the hair sampled) was found to be a factor of more than 2 larger than that in the control population (+/- 58% vs +/- 26%). A strong relationship (R = 0.656; P < .01) between log H-Cr(pr) and log fasting plasma Cr was observed in the diabetic group (n = 20). The mean fasting plasma Cr value of this group was 0.41 +/- 0.10 microg Cr per liter. No correlation between H-Cr(av.) and duration of diabetes was observed. A strong positive association was observed in the control population between H-Cr(pr) and fasting plasma insulin (n = 22; R = 0.6157; P < .01), and H-Cr(pr) and fasting plasma glucose (n = 24; R = 0.4118; P < .05), which is indicative of the interrelation of these parameters. In the control population, H-Cr(av) showed a slight decrease with age (n = 54; R = 0.2691; P < .05), which is assumed to be the result of increased insulin resistance caused by various age-associated factors including Cr deficiency. None of the above relationships was significant in the diabetic group. Evidence is presented that justifies the assumption that the longitudinal H-Cr profile resembles the variation in Cr metabolic rate over the time span of growing hair, which is not appreciably affected by external contamination. This suggests that glucose intolerance (type 2 diabetes mellitus) is an important factor that disturbs Cr metabolism.
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
Dimethylamine, anhydrous, ≥99%
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Sigma-Aldrich
Pyruvaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Dodecanedioic acid, 99%
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Supelco
1-Butanol, analytical standard
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetone, analytical standard
Sigma-Aldrich
1-Butanol, BioRenewable, ACS reagent, ≥99.4%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Sigma-Aldrich
3-Hydroxybutyric acid, 95%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%