Merck
  • Gas-liquid chromatographic determination of galactitol in amniotic fluid for possible use in prenatal diagnosis of galactosaemia.
Gas-liquid chromatographic determination of galactitol in amniotic fluid for possible use in prenatal diagnosis of galactosaemia.
Clinica chimica acta; international journal of clinical chemistry (1981-02-19)
J T Allen, J B Holton, M G Gillett
ABSTRACT
A gas liquid chromatographic method for the estimation of galactitol in amniotic fluid from pregnancies at risk for galactosaemia is described. The method is based on the almost complete removal of glucose from the amniotic fluid by ion exchange, and the subsequent chromatography of galactitol as its hexaacetate.
Sigma-Aldrich
Dulcitol, ≥99% (GC)