Merck
  • High mobility, low voltage operating C(60) based n-type organic field effect transistors.
High mobility, low voltage operating C(60) based n-type organic field effect transistors.
Synthetic metals (2011-11-04)
G Schwabegger, Mujeeb Ullah, M Irimia-Vladu, M Baumgartner, Y Kanbur, R Ahmed, P Stadler, S Bauer, N S Sariciftci, H Sitter
ABSTRACT
We report on C(60) based organic field effect transistors (OFETs) that are well optimized for low voltage operation. By replacing commonly used dielectric layers by thin parylene films or by utilizing different organic materials like divinyltetramethyldisiloxane-bis(benzocyclo-butene) (BCB), low density polyethylene (PE) or adenine in combination with aluminum oxide (AlOx) to form a bilayer gate dielectric, it was possible to significantly increase the capacitance per unit area (up to two orders of magnitude). The assembly of metal-oxide and organic passivation layer combines the properties of the high dielectric constant of the metal oxide and the good organic-organic interface between semiconductor and insulator provided by a thin capping layer on top of the AlOx film. This results in OFETs that operate with voltages lower than 500 mV, while exhibiting field effect mobilities exceeding 3 cm(2) V(-1) s(-1).
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Medium density
Supelco
Polyethylene, analytical standard, suitable for gel permeation chromatography (GPC), 2,000
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000