Merck
  • Cyclophilin A differentially activates monocytes and endothelial cells: role of purity, activity, and endotoxin contamination in commercial preparations.
Cyclophilin A differentially activates monocytes and endothelial cells: role of purity, activity, and endotoxin contamination in commercial preparations.
Atherosclerosis (2007-10-09)
Sravan K Payeli, Cordelia Schiene-Fischer, Jan Steffel, Giovanni G Camici, Izabela Rozenberg, Thomas F Lüscher, Felix C Tanner
ABSTRACT
Cyclophilin A (CyPA) is a cytoplasmic protein secreted under inflammatory conditions. Extracellular CyPA is detected in atherosclerotic plaques and has been observed to activate endothelial cells as well as monocytes. Commercially available recombinant CyPA-induced expression of tissue factor (TF) and vascular cell adhesion molecule-1 (VCAM-1) in human aortic endothelial cells (HAEC). However, CyPA from commercial sources contained lipopolysaccharide at concentrations up to 18.9 ng/ml; moreover, it exhibited low purity as determined by protein spectrum analysis and low activity as assessed by peptidyl prolyl cis-trans isomerase (PPIase) assay. An in-house preparation of pure, active, and uncontaminated CyPA failed to induce endothelial TF or VCAM-1 expression; moreover, it was not chemotactic for HAEC. In contrast, such CyPA exhibited potent chemotactic activity on monocytic THP-1 cells, with a maximal effect on migration occurring at a concentration of 5.5 x 10(-9)mol/l. Pretreatment of CyPA with cyclosporine A prevented its effect on THP-1 cell migration; similarly, PPIase-deficient mutant CyPA protein did not induce migration of these cells. In-house prepared CyPA induced the release of Il-6, but not TNF-alpha, from THP-1 cells. Commercially available CyPA exhibits low purity and activity and may be contaminated by endotoxin. Pure, active, and uncontaminated CyPA does not induce endothelial TF or VCAM-1 expression; instead, it acts as a potent monocyte chemoattractant and induces monocyte Il-6 release, implying a role for extracellular CyPA in the pathogenesis of atherosclerosis via activation of monocytes rather than endothelial cells.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Roche
Cytotoxicity Detection Kit (LDH), suitable for protein quantification, suitable for cell analysis, detection, sufficient for ≤2,000 tests
Sigma-Aldrich
Cyclophilin A human, ≥95% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous solution