Merck
Genotoxicity in herring gulls (Larus argentatus) in Sweden and Iceland.
Mutation research (2010-07-21)
Halldora Skarphedinsdottir, Gunnar Thor Hallgrimsson, Tomas Hansson, Per-Ake Hägerroth, Birgitta Liewenborg, Ulla Tjärnlund, Gun Akerman, Janina Barsiene, Lennart Balk
ABSTRACT
Adult and young herring gulls (Larus argentatus) in Sweden and Iceland were investigated with respect to DNA adducts, analysed with the nuclease-P1 version of the (32)P-postlabelling method, and micronucleated erythrocytes. Three important aims were: (1) to estimate the degree of exposure to genotoxic environmental pollutants in the Baltic Sea area and Iceland, (2) to evaluate the utility of the investigated biomarkers in birds, and (3) to investigate if there was any relationship between genotoxic effects and thiamine deficiency. The results demonstrate that both Swedish and Icelandic herring gulls are exposed to genotoxic pollution. Urban specimens have higher levels of DNA adducts than rural specimens, but background exposure to genotoxic environmental pollutants, such as PAHs, is also significant. In the herring gull the general level of DNA adducts in the liver seems to be higher than in fish. DNA adducts were most abundant in the liver, followed by the kidney, intestinal mucosa, and whole blood, in decreasing order. The frequency of micronucleated erythrocytes was probably slightly elevated in all the investigated sites, reflecting a significant background exposure. The level of DNA adducts was unrelated to the frequency of micronucleated erythrocytes, and both these variables were unrelated to symptoms of thiamine deficiency. The investigation confirmed the utility of DNA adducts, and probably also micronucleated erythrocytes, as biomarkers of genotoxicity in birds.
Sigma-Aldrich
Phosphodiesterase II from bovine spleen, lyophilized powder, ≥5.0 units/mg protein