Merck
  • Effect of ionising radiation on polyphenolic content and antioxidant potential of parathion-treated sage (Salvia officinalis) leaves.
Effect of ionising radiation on polyphenolic content and antioxidant potential of parathion-treated sage (Salvia officinalis) leaves.
Food chemistry (2013-06-25)
Issam Ben Salem, Sana Fekih, Haitham Sghaier, Mehrez Bousselmi, Mouldi Saidi, Ahmed Landoulsi, Sami Fattouch
ABSTRACT
The γ-irradiation effects on polyphenolic content and antioxidant capacity of parathion-pretreated leaves of Salvia officinalis plant were investigated. The analysis of phenolic extracts of sage without parathion showed that irradiation decreased polyphenolic content significantly (p<0.05) by 30% and 45% at 2 and 4kGy, respectively, compared to non-irradiated samples. The same trend was observed for the Trolox equivalent antioxidant capacity (TEAC), as assessed by the anionic DPPH and cationic ABTS radical-scavenging assays. The antioxidant potential decreased significantly (p<0.01) at 2 and 4kGy, by 11-20% and 40-44%, respectively. The results obtained with a pure chlorogenic acid solution confirmed the degradation of phenols; however, its TEAC was significantly (p<0.01) increased following irradiation. Degradation products of parathion formed by irradiation seem to protect against a decline of antioxidant capacity and reduce polyphenolic loss. Ionising radiation was found to be useful in breaking down pesticide residues without inducing significant losses in polyphenols.
Product Description
Supelco
Parathion solution, 100 μg/mL in cyclohexane, PESTANAL®, analytical standard
Supelco
Parathion, PESTANAL®, analytical standard