Merck
  • Which agonist properties are important for the activation of 5-HT3A receptors?
Which agonist properties are important for the activation of 5-HT3A receptors?
Biochimica et biophysica acta (2013-06-25)
M F Meiboom, M Barann, S Witten, K Groeneveld, B W Urban
ABSTRACT
Why do anesthetics not activate excitatory ligand-gated ion channels such as 5-HT3 receptors in contrast to inhibitory ligand-gated ion channels? This study examines the actions of structural closely-related 5-HT derivatives and 5-HT constituent parts on 5-HT3A receptors with the aim of finding simpler if not minimal agonists and thus determining requirements for successful agonist action. Responses to 5-HT derivatives of human 5-HT3A receptors stably expressed in HEK 293 cells have been examined with the patch-clamp technique in the outside-out configuration combined with a fast solution exchange system. Phenol, pyrrole and alkyl amines, constituents of 5-HT, even at high concentrations, cannot activate 5-HT3A receptors but they can inhibit them. To date, tyramines are the smallest known agonists. However, an aromatic ring is not required for activation as acetylcholine is also an agonist of similar strength. Simultaneous interactions of adequate strength at two separate subsites within the 5-HT binding domain appear to be essential for successful agonist function. Anesthetics either fail to achieve this or the activation they produce is so weak that it is masked by a comparatively very strong inhibition.
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)