Merck
  • Enhanced NIR downconversion luminescence by precipitating nano Ca5(PO4)3F crystals in Eu2+-Yb3+ co-doped glass.
Enhanced NIR downconversion luminescence by precipitating nano Ca5(PO4)3F crystals in Eu2+-Yb3+ co-doped glass.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-07-03)
Chen Li, Zhiguo Song, Yongjin Li, Kai Lou, Jianbei Qiu, Zhengwen Yang, Zhaoyi Yin, Xue Wang, Qi Wang, Ronghua Wan
ABSTRACT
Eu(2+)-Yb(3+) co-doped transparent glass-ceramic containing nano-Ca5(PO4)3F (FAP) was prepared in reducing atmosphere. XRD and TEM analysis indicated that nano-FAP about 40 nm precipitated homogeneously in glass matrix after heat treatment. Confirmed by spectroscopy measurements, the crystal-like absorption and emission of Eu(2+) indicated the partition of Eu(2+) into FAP nanocrystals in glass ceramic. NIR emission due to the transition (2)F5/2→(2)F7/2 of Yb(3+) ions (about 980-1100 nm) was observed from glasses under ultraviolet excitation, ascribed to downconversion from Eu(2+) to Yb(3+), which can be enhanced by precipitating nano-FAP crystals. The results indicated that Eu(2+)-Yb(3+) co-doped glass-ceramic embedding with nano-FAP is a promising candidate as downconversion materials for enhancing conversion efficiency of solar cells.
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), ≥97%, synthetic
Sigma-Aldrich
Ytterbium, powder, ≥99.9% trace rare earth metals basis
Sigma-Aldrich
Hydroxyapatite, synthetic, 99.8% trace metals basis (excludes Mg)
Sigma-Aldrich
Hydroxyapatite, nanoparticles, dispersion, 10 wt. % in H2O, <200 nm particle size (BET)
Sigma-Aldrich
Calcium phosphate tribasic, BioReagent, suitable for plant cell culture, powder
Sigma-Aldrich
Calcium phosphate tribasic, 34.0-40.0% Ca basis
Sigma-Aldrich
Hydroxyapatite, powder, synthetic
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), contains 5 wt. % silica as dopant, synthetic