Merck
  • Calcium supplementation during sepsis exacerbates organ failure and mortality via calcium/calmodulin-dependent protein kinase kinase signaling.
Calcium supplementation during sepsis exacerbates organ failure and mortality via calcium/calmodulin-dependent protein kinase kinase signaling.
Critical care medicine (2013-07-28)
Richard D Collage, Gina M Howell, Xianghong Zhang, Jennifer L Stripay, Janet S Lee, Derek C Angus, Matthew R Rosengart
ABSTRACT
Calcium plays an essential role in nearly all cellular processes. As such, cellular and systemic calcium concentrations are tightly regulated. During sepsis, derangements in such tight regulation frequently occur, and treating hypocalcemia with parenteral calcium administration remains the current practice guideline. We investigated whether calcium administration worsens mortality and organ dysfunction using an experimental murine model of sepsis and explored the mechanistic role of the family of calcium/calmodulin-dependent protein kinases in mediating these physiological effects. To highlight the biological relevance of these observations, we conducted a translational study of the association between calcium administration, organ dysfunction, and mortality among a cohort of critically ill septic ICU patients. Prospective, randomized controlled experimental murine study and observational clinical cohort analysis. University research laboratory and eight ICUs at a tertiary care center. A cohort of 870 septic ICU patients. C57Bl/6 and CaMKK mice. Mice underwent cecal ligation and puncture polymicrobial sepsis and were administered with calcium chloride (0.25 or 0.25 mg/kg) or normal saline. Administering calcium chloride to septic C57Bl/6 mice heightened systemic inflammation and vascular leak, exacerbated hepatic and renal dysfunction, and increased mortality. These events were significantly attenuated in CaMKK mice. In a risk-adjusted analysis of septic patients, calcium administration was associated with an increased risk of death, odds ratio 1.92 (95% CI, 1.00-3.68; p = 0.049), a significant increase in the risk of renal dysfunction, odds ratio 4.74 (95% CI, 2.48-9.08; p < 0.001), and a significant reduction in ventilator-free days, mean decrease 3.29 days (0.50-6.08 days; p = 0.02). Derangements in calcium homeostasis occur during sepsis that is sensitive to calcium administration. This altered calcium signaling, transduced by the calmodulin-dependent protein kinase kinase cascade, mediates heightened inflammation and vascular leak that culminates in elevated organ dysfunction and mortality. In the clinical management of septic patients, calcium supplementation provides no benefit and may impose harm.
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride dihydrate, puriss., meets analytical specification of Ph. Eur., USP, FCC, E509, 99-103%, ≤0.0001% Al
Sigma-Aldrich
Calcium chloride dihydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Calcium chloride dihydrate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium chloride dihydrate, BioUltra, Molecular Biology, ≥99.5% (KT)
Sigma-Aldrich
Calcium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Calcium chloride dihydrate, Molecular Biology, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, ReagentPlus®, ≥99.0%