Merck
  • Microabrasion using 18% hydrochloric acid and 37% phosphoric acid in various degrees of fluorosis - an in vivo comparision.
Microabrasion using 18% hydrochloric acid and 37% phosphoric acid in various degrees of fluorosis - an in vivo comparision.
The European journal of esthetic dentistry : official journal of the European Academy of Esthetic Dentistry (2013-08-21)
Soumya Sinha, Kiran Kumar Vorse, Hina Noorani, Shivprakash Pujari Kumaraswamy, Siddhartha Varma, Haragopal Surappaneni
ABSTRACT
The aim of this study was to assess the efficacy of 18% hydrochloric acid and 37% phosphoric acid by an in vivo comparison. Sixty fluorotic permanent maxillary central incisors from 30 patients were divided into 3 categories. The teeth received 5 seconds (mild fluorosis), 20 seconds (moderate fluorosis) and 30 seconds (severe fluorosis) application of 18% hydrochloric acid on 11 and 37% phosphoric acid on 21. Standardized intraoral photographies were taken immediately before, after, and one month after treatment. Vinyl polysiloxane impression of the patient were made before and after the treatment. A scanning electron microscopic (SEM) evaluation was carried out on the models to judge the surface alterations. Wilcoxon and Mann-Whitney tests were used to verify the hypothesis. A statistically significant result was obtained in the reduction of white spot opacities, intensity of stains and the total area occupied by the stains in mild and moderate fluorosis teeth. Results of severe fluorosis had an unpredictable outcome. An SEM evaluation revealed good improvement in the surface texture of mild and moderate fluorosis teeth. Teeth with severe fluorosis showed only a slight improvement. A microabrasion procedure is effective for treating mild and moderate fluorosis cases.
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Supelco
Hydrogen chloride - 1-butanol solution, ~3 M in 1-butanol, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
Sigma-Aldrich
Phosphoric acid, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Phosphoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Phosphoric acid, puriss., ≥99%
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10