Merck
  • [Bone alkaline phosphatase and N-MID osteocalcin in monitoring of osteoporosis treatment with recombinant human parathyroid hormone 1-34].
[Bone alkaline phosphatase and N-MID osteocalcin in monitoring of osteoporosis treatment with recombinant human parathyroid hormone 1-34].
Zhejiang da xue xue bao. Yi xue ban = Journal of Zhejiang University. Medical sciences (2013-10-30)
Xiu-Mei Weng, Jian-Ping Pan
ABSTRACT
To evaluate the application of bone turnover markers bone alkaline phosphatase (BALP) and N-MID osteocalcin (N-MID) in monitoring of osteoporosis treatment with recombined parathyroid hormone 1-34 (rhPTH1-34). The bone mineral density (BMD) of the lumbar spine L2-L4 and the proximal femur were examined by dual energy X-ray absorptiometry (DXA) before and 6 and 12 months after rhPTH 1-34 treatment. Meanwhile, serum levels of BALP and N-MID were detected by electro-chemiluminescence assay. Six months after rhPTH 1-34 treatment, the BMD of proximal femur remained unchanged, and the BMD of the lumbar L2-L4 spine increased from 0.753 g/cm(2) to 0.781 g/cm(2) (P<0.05); while serum levels of N-MID increased from 15.46 ng/ml to 27.07 ng/ml(P<0.01), BALP from 14.05 μg/ml to 24.31 μg/ml(P<0.01). Twelve months after drug administration, no significant changes were observed in BMD of proximal femur, and the BMD of the lumbar spine L2-L4 increased from 0.753 g/cm(2) to 0.807 g/cm(2)(P<0.01) while serum levels of N-MID and BALP increased from 15.46 ng/ml and 14.05μg/ml to 49.38 ng/ml and 33.99 μg/ml, respectively (both P<0.01). Serum levels of BALP and N-MID are more sensitive than BMD. Combination of two methods may provide better indicators for monitoring of osteoporosis treatment.
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from calf intestinal mucosa, suitable for enzyme immunoassay, solution (clear, colorless), ~2500 U/mg protein (~10 mg/ml)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein