Merck
  • Osmolyte effects on the self-association of concanavalin A: testing theoretical models.
Osmolyte effects on the self-association of concanavalin A: testing theoretical models.
Biochemistry (2013-11-13)
Thomas R Silvers, Jeffrey K Myers
ABSTRACT
The formation and stability of protein-protein interfaces are of obvious biological importance. While a large body of literature exists describing the effect of osmolytes on protein folding, very few studies address the effect of osmolytes on protein association and binding. The plant lectin concanavalin A (ConA), which undergoes a reversible tetramer-to-dimer equilibrium as a function of pH, was used as a model system to investigate the influence of nine osmolytes on protein self-association. The stabilizing or destabilizing impacts of the osmolytes were evaluated from pH titrations combined with circular dichroism spectroscopy. Relative to the dimer, trimethylamine N-oxide, betaine, proline, sarcosine, sorbitol, sucrose, and trehalose all stabilized the ConA tetramer to varying extents. Glycerol had a negligible effect, and urea destabilized the tetramer. From multiple titrations in different osmolyte concentrations, an m-value (a thermodynamic parameter describing the change in the association free energy per molar of osmolyte) was determined for each osmolyte. Experimental m-values were compared with those calculated using two theoretical models. The Tanford transfer model, with transfer free energies determined by Bolen and co-workers, failed to accurately predict the m-values in most cases. A model developed by Record and co-workers, currently applicable only to urea, betaine, and proline, more accurately predicted our experimental m-values, but significant discrepancies remained. Further theoretical work is needed to develop a thermodynamic model to predict the effect of osmolytes on protein-protein interfaces, and further experimental work is needed to determine if there is a general stabilization by osmolytes of such interfaces.
Product Description
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), Type IV-S, lyophilized powder, aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), Type VI, lyophilized powder
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), Molecular Biology
SAFC
L-Proline
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), Type IV-S, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), Type IV, lyophilized powder
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sigma-Aldrich
Sarcosine, crystallized, ≥98.0% (T)
Sigma-Aldrich
Sarcosine, 98%
Sigma-Aldrich
Sarcosine, BioXtra
Supelco
L-Proline, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)