Merck
  • Doxorubicin has in vivo toxicological effects on ex vivo cultured mesenchymal stem cells.
Doxorubicin has in vivo toxicological effects on ex vivo cultured mesenchymal stem cells.
Toxicology letters (2013-12-03)
Maira Souza Oliveira, Juliana Lott Carvalho, Ana Carolina De Angelis Campos, Dawidson Assis Gomes, Alfredo Miranda de Goes, Marília Martins Melo
ABSTRACT
Doxorubicin (dox) is an effective chemotherapeutic agent that leads to cardiotoxicity. An alternative treatment for dox-cardiotoxicity is autologous mesenchymal stem cells (MSCs) transplantation. It remains unclear if dox has deleterious effects on MSCs from subjects under chemotherapy, therefore this study aimed to evaluate dox in vivo toxicological effects on ex vivo cultured MSCs, inferring whether autologous transplantation may be an alternative treatment in patients who are exposed to the drug. Wistar rats received either dox or saline. Following treatments, animals were sacrificed and bone marrow MSCs were isolated, characterized for cell surface markers and assessed according to their viability, alkaline phosphatase production, and proliferation kinetics. Moreover, MSCs were primed to cardiac differentiation and troponin T and connexin 43 expressions were evaluated. Compared to control, undifferentiated MSCs from dox group kept the pattern for surface marker and had similar viability results. In contrast, they showed lower alkaline phosphatase production, proliferation rate, and connexin 43 expression. Primed MSCs from dox group showed lower troponin T levels. It was demonstrated a toxic effect of dox in host MSCs. This result renders the possibility of autologous MSCs transplantation to treat dox-cardiotoxicity, which could be a non-suitable option for subjects receiving such antineoplastic agent.
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from calf intestinal mucosa, suitable for enzyme immunoassay, solution (clear, colorless), ~2500 U/mg protein (~10 mg/ml)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host
Doxorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)