Merck
  • [Substrate specificity of Cryptococcus albidus and Eupenicillium erubescens alpha-L-rhamnosidases].
[Substrate specificity of Cryptococcus albidus and Eupenicillium erubescens alpha-L-rhamnosidases].
Ukrains'kyi biokhimichnyi zhurnal (1999 ) (2014-02-01)
O V Hudzenko, L D Varbanets'
ABSTRACT
The substrate specificity of Cryptococcus albidus and Eupenicillium erubescens alpha-L-rhamnosidases has been investigated. It is shown that the enzymes are able to act on synthetic and natural substrates, such as naringin, neohesperidin. alpha-L-Rhamnosidases hydrolysed the latter ones very efficiently, in this case E. erubescens enzyme was characterized by higher values of V(max) in comparison with the enzyme of C. albidus. However the C. albidus alpha-L-rhamnosidase showed greater affinity for naringin and neohesperidin than the enzyme of E. erubescens (K(m) 0.77 and 3.3 mM and 5.0 and 3.0 mM, respectively). As regards the synthetic derivatives of monosaccharides, both enzymes exhibited narrow specificity for glycon: E. erubescens alpha-L-rhamnosidase--only to the p-nitrophenyl-alpha-L-rhamnopiranoside (K(m) 1.0 mM, V(max) 120 micromol/min/mg protein), and C. albidus--to p-nitrophenyl-alpha-D-glucopyranoside (K(m) 10 mM, V(max) 5 micromol/min/mg protein). Thus, it was found that the enzyme preparations of E. erubescens and C. albidus are differed by their substrate specificity. The ability of E. erubescens and C. albidus alpha-L-rhamnosidases to hydrolyse natural substrates: naringin and neohesperidin, evidences for their specificity for alpha-1,2-linked L-rhamnose. Based on these data, we can predict the use of E. erubescens and C. albidus alpha-L-rhamnosidases in various industries, food industry in particular. This is also confirmed by the fact that the investigated alpha-L-rhamnosidases were stable at 20% concentration of ethanol and 500 mM glucose in the reaction mixture.