Merck
Priapism in a patient with unstable hemoglobin: hemoglobin Köln.
American journal of hematology (2003-09-02)
Valérie Andrieu, Olivier Dumonceau, Marie-José Grange
ABSTRACT
We report a case of severe priapism occurring in a patient with unstable hemoglobin, hemoglobin Köln, and underline several factors that may have contributed to this complication: abnormal plasticity of red cells, splenectomy, and cytomegalovirus infection. Since emergency treatment may prevent impotence, patients and parents should be educated about this complication.
Product Description
Etilefrine hydrochloride, European Pharmacopoeia (EP) Reference Standard