Merck
  • Improved post-prandial ghrelin response by nateglinide or acarbose therapy contributes to glucose stability in Type 2 diabetic patients.
Improved post-prandial ghrelin response by nateglinide or acarbose therapy contributes to glucose stability in Type 2 diabetic patients.
Journal of endocrinological investigation (2013-01-18)
F Zheng, X Yin, W Lu, J Zhou, H Yuan, H Li
ABSTRACT
Recent studies highlight an important role of ghrelin in glucose homeostasis, while the association between ghrelin regulation and glucose fluctuation is unclear. We compared the effects of two postprandial hypoglycemic agents on ghrelin response and determined the contribution of ghrelin response to glucose stability in Type 2 diabetic (T2DM) patients. Forty newly- diagnosed T2DM patients were randomly allocated to receive nateglinide or acarbose for 4 weeks, with twenty body mass index (BMI)-matched normoglycemic subjects as controls. Mean glucose values and daily average glucose excursion were assessed using continuous glucose monitoring system. Serum ghrelin levels were determined by enzyme-linked immunosorbent assay. T2DM patients had similar fasting ghrelin levels (p=0.546), while their postprandial ghrelin suppressions at 30 min and 120 min were reduced as compared to BMI-matched normoglycemic controls (p<0.01). Both nateglinide and acarbose increased post-prandial ghrelin suppression at 120 min and reduced ghrelin area under the curve (AUCGHRL) (p<0.05), while only nateglinide increased postprandial ghrelin suppression at 30 min (p<0.01), which was positively correlated with the increased early-phase insulin secretion by 4 weeks of nateglinide therapy (r=0.48, p=0.05). The decrease in AUCGHRL was positively correlated with the decrease in daily average glucose excursion and mean glucose values either by 4 weeks of nateglinide or acarbose therapy (p<0.05). Both nateglinide and acarbose increase post-prandial ghrelin suppression. Improved ghrelin regulation is most likely to play a role in glucose stability in T2DM patients with nateglinide or acarbose therapy.
Product Description
N-[(cis-4-Isopropylcyclohexyl)carbonyl]-D-phenylalanine, European Pharmacopoeia (EP) Reference Standard
USP
Nateglinide Related Compound C, United States Pharmacopeia (USP) Reference Standard