Merck
Rickettsial fever presenting with isolated third nerve palsy.
The Journal of the Association of Physicians of India (2014-01-30)
Anjali Rajadhyaksha, Sanat Phatak, Nilesh Nolkha, Yasmeen Pathan, Archana Sonawale
ABSTRACT
Rickettsial fevers are known to have neurological involvement, mostly in the form of meningoencephalitis. Focal neurodeficits, including isolated cranial nerve palsies have been rarely reported. We hereby report a case of a 25 year old man who presented to us with high grade fever caused by rickettsia and left sided partial third cranial nerve palsy. He responded to doxycycline.