Merck
  • Sensitive bioanalysis: combining single-molecule spectroscopy with mono-labeled self-quenching probes.
Sensitive bioanalysis: combining single-molecule spectroscopy with mono-labeled self-quenching probes.
Analytical and bioanalytical chemistry (2007-06-15)
Nicole Marmé, Jens-Peter Knemeyer
ABSTRACT
Fluorescence single-molecule spectroscopy is an appropriate tool for modern bioanalysis. This technique enables the development of ultra sensitive assays, especially when combined with self-quenching probes. In this review we report novel DNA, enzyme, and antibody assays based on mono-labeled fluorescent probes that are quenched by photoinduced electron transfer (PET).
Sigma-Aldrich
Atto 680 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 655-Biotin, BioReagent, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Atto 655, BioReagent, suitable for fluorescence, ≥85% (HPLC)
Sigma-Aldrich
Atto 655-maleimide, BioReagent, suitable for fluorescence, ≥90% (coupling rate)
Sigma-Aldrich
Atto 655 NHS ester, BioReagent, suitable for fluorescence