Merck
  • Using the yeast two-hybrid system to identify protein-protein interactions.
Using the yeast two-hybrid system to identify protein-protein interactions.
Methods in molecular biology (Clifton, N.J.) (2013-10-19)
Edgar Rodríguez-Negrete, Eduardo R Bejarano, Araceli G Castillo
ABSTRACT
The yeast two-hybrid system is currently one of the most standardized protein interaction mapping techniques. The rationale of the yeast two-hybrid system relies on the physical separation of the DNA-binding domain from the transcriptional activation domain of several transcription factors. The protein of interest (bait) is fused to a DNA-binding domain, and complementary DNA (cDNA) library-encoded proteins are fused to a transcriptional activation domain. When a protein encoded by the cDNA library binds to the bait, both activities of the transcription factor are rejoined resulting in transcription from a reporter gene. Here, we describe protocols to test interactions between two individual proteins and to look for novel interacting partners by screening a single protein or domain against a library of other proteins using a GAL4 based yeast two-hybrid system.
Sigma-Aldrich
β-Galactosidase from Aspergillus oryzae, ≥8.0 units/mg solid
Sigma-Aldrich
β-Galactosidase from Escherichia coli, suitable for enzyme immunoassay, lyophilized, powder, ~140 U/mg
Sigma-Aldrich
β-Galactosidase from bovine testes, ammonium sulfate suspension, 1.0-3.0 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
β-Galactosidase from Escherichia coli, Grade VI, lyophilized powder, ≥250 units/mg protein
Sigma-Aldrich
β-Galactosidase from bovine liver, Grade III, lyophilized powder, ≥0.15 units/mg protein
Sigma-Aldrich
β-Galactosidase from Escherichia coli, lyophilized powder, ≥500 units/mg protein
Sigma-Aldrich
β-Galactosidase from Escherichia coli, Grade VIII, lyophilized powder, ≥500 units/mg protein
Sigma-Aldrich
β-Galactosidase from Escherichia coli, aqueous glycerol suspension, ≥500 units/mg protein (biuret)