Merck
  • Evaluation of first-pass cytochrome P4503A (CYP3A) and P-glycoprotein activities using felodipine and hesperetin in combination in Wistar rats and everted rat gut sacs in vitro.
Evaluation of first-pass cytochrome P4503A (CYP3A) and P-glycoprotein activities using felodipine and hesperetin in combination in Wistar rats and everted rat gut sacs in vitro.
Phytotherapy research : PTR (2013-07-25)
V Sridhar, M Surya Sandeep, P Ravindra Babu, K Naveen Babu
ABSTRACT
The effects of hesperetin on the pharmacokinetics and the role of P-glycoprotein (P-gp) in the transport of felodipine were investigated in rats and in vitro. Felodipine was administered orally (10 mg/kg) without or with hesperetin (25, 50 and 100 mg/kg) to rats for 15 consecutive days. Blood samples were collected at different time intervals on 1(st) day in single dose pharmacokinetic study (SDS) and on 15(th) day in multiple dose pharmacokinetic study (MDS). The area under the plasma concentration-time curve (AUC0-∞ ) and the peak plasma concentration (Cmax ) of felodipine were dose-dependently increased in SDS and MDS with hesperetin compared to control ( p < 0.001). The half-life (t1/2 ) and mean residence time was longer than the control group in both studies. The role of P-gp determined using everted rat gut sacs in vitro by incubating felodipine with or without hesperetin and verapamil (typical P-gp and CYP3A4 inhibitor). The in vitro experiments revealed that the verapamil and hesperetin increased the intestinal absorption of felodipine (p < 0.01). Concurrent use of hesperetin dramatically altered the pharmacokinetics of felodipine leading to an increase in systemic exposure. The likely mechanism is inhibition of CYP3A4-mediated first-pass metabolism and P-gp in the intestine and the liver.
Sigma-Aldrich
Felodipine, solid
Supelco
(±)-Verapamil hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder
Supelco
Verapamil hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Felodipine, European Pharmacopoeia (EP) Reference Standard
USP
Verapamil hydrochloride, United States Pharmacopeia (USP) Reference Standard
Verapamil hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hesperidin, ≥80% (HPLC)
Supelco
Hesperidin, analytical standard
Hesperidin, European Pharmacopoeia (EP) Reference Standard