Merck
  • Neuroprotective role of hydralazine in rat spinal cord injury-attenuation of acrolein-mediated damage.
Neuroprotective role of hydralazine in rat spinal cord injury-attenuation of acrolein-mediated damage.
Journal of neurochemistry (2013-11-30)
Jonghyuck Park, Lingxing Zheng, Andrew Marquis, Michael Walls, Brad Duerstock, Amber Pond, Sasha Vega-Alvarez, He Wang, Zheng Ouyang, Riyi Shi
ABSTRACT
Acrolein, an α,β-unsaturated aldehyde and a reactive product of lipid peroxidation, has been suggested as a key factor in neural post-traumatic secondary injury in spinal cord injury (SCI), mainly based on in vitro and ex vivo evidence. Here, we demonstrate an increase of acrolein up to 300%; the elevation lasted at least 2 weeks in a rat SCI model. More importantly, hydralazine, a known acrolein scavenger can provide neuroprotection when applied systemically. Besides effectively reducing acrolein, hydralazine treatment also resulted in significant amelioration of tissue damage, motor deficits, and neuropathic pain. This effect was further supported by demonstrating the ability of hydralazine to reach spinal cord tissue at a therapeutic level following intraperitoneal application. This suggests that hydralazine is an effective neuroprotective agent not only in vitro, but in a live animal model of SCI as well. Finally, the role of acrolein in SCI was further validated by the fact that acrolein injection into the spinal cord caused significant SCI-like tissue damage and motor deficits. Taken together, available evidence strongly suggests a critical causal role of acrolein in the pathogenesis of spinal cord trauma. Since acrolein has been linked to a variety of illness and conditions, we believe that acrolein-scavenging measures have the potential to be expanded significantly ensuring a broad impact on human health.
Sigma-Aldrich
Hydralazine hydrochloride
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Hydralazine hydrochloride, European Pharmacopoeia (EP) Reference Standard