Merck
  • DDTs in mothers' milk, placenta and hair, and health risk assessment for infants at two coastal and inland cities in China.
DDTs in mothers' milk, placenta and hair, and health risk assessment for infants at two coastal and inland cities in China.
Environment international (2014-01-30)
Yu Bon Man, Janet Kit Yan Chan, Hong Sheng Wang, Sheng Chun Wu, Ming Hung Wong
ABSTRACT
This study is a one of the very few investigating the dichloro-diphenyl-trichloroethanes (DDTs) (summation of o,p'-DDE, p,p'-DDE, o,p'-DDD, p,p'-DDD, o,p'-DDT, and p,p'-DDT) in multiple human matrices in mothers' milk, placenta and hair collected from residents from two coastal cities: Guiyu (GY) and Taizhou (TZ) and one inland city: Lin'an (LA). TZ (milk: 360±319ng/g lipid wt.) showed significantly higher concentrations of DDTs than those from LA (milk: 190±131ng/g lipid wt.), whereas, concentrations of DDTs in GY (milk: 305±109ng/g lipid wt.) were in between TZ and LA. In addition, levels of DDTs in the human tissues from TZ (placenta: 122±109ng/g lipid wt.; hair: 79.9±215ng/g dry wt.) were significantly higher than those from Lin'an (placenta: 49.2±30.2ng/g lipid wt.; hair: 10.8±7.09ng/g dry wt.). The above concentrations of DDTs in milk exceeded the Codex Maximum Residue Limits/Extraneous Maximum Residue Limits for milk (20ng/g lipid wt. whole milk), indicating that the human milk samples were grossly polluted. The present study revealed that human specimens collected from the coastal city (TZ) were more contaminated with inland one (LA), based on the levels of DDTs contained in samples which may be due to the higher dietary exposure to DDTs via consumption of contaminated seafood. The estimated daily intakes of DDTs by GY, TZ and LA infants were 1.69±1.86, 1.48±0.79, and 0.95±0.73μg/kg body wt./day, respectively which did not exceed 10μg/kg body wt./day, the provisional tolerable daily intake proposed by the Food and Agriculture Organization/World Health Organization.
Product Description
Supelco
4,4′-DDT solution, certified reference material, 5000 μg/mL in methanol
Supelco
4,4′-DDT, PESTANAL®, analytical standard