Merck
  • Mercury in the fairy-ring of Gymnopus erythropus (Pers.) and Marasmius dryophilus (Bull.) P. Karst. mushrooms from the Gongga Mountain, Eastern Tibetan Plateau.
Mercury in the fairy-ring of Gymnopus erythropus (Pers.) and Marasmius dryophilus (Bull.) P. Karst. mushrooms from the Gongga Mountain, Eastern Tibetan Plateau.
Ecotoxicology and environmental safety (2014-03-19)
Jerzy Falandysz, Anna Dryżałowska, Martyna Saba, Jipeng Wang, Dan Zhang
ABSTRACT
Gongga Mountain or Minya Konka, like the Himalayan Dimension Mountains, has its own microclimate and a 'circum-polar' climate and hence is sensitive to contamination by persistent pollutants that are trapped by cold temperature and wet precipitation. Elemental mercury (Hg) as vapour easy diffuses into the atmosphere and the rate of Hg deposition from global fallout is dependent on locally ambient temperature and precipitation. We investigated the accumulation and distribution of total Hg in two species of mushrooms, Gymnopus erythropus and Marasmius dryophilus, which grew on Gongga Mountain. The fruiting bodies were collected at a height of 2946m above see level. Both species efficiently accumulated Hg. The median values for caps of M. dryophilus and G. erythropus were 1.168 and 3.078, and for stipes 0.573 and 1.636mg/kg dry matter, respectively, and in the beneath litter and soil were 0.13 and 0.15mg/kg dry matter. The Hg contents of the caps of M. dryophilus and the beneath litter and soils from pristine Himalayan forest of 1.168, 0.132 and 0.116mg/kg dry matter (respectively) is high compared to values reported for similar species and soils from background areas in Poland -0.58-0.70 and 0.047-0.048mg/kg dry matter. The absence of industrial activities, urbanization and Hg ore deposits at Gongga Mountain suggests that long-range atmospheric transport and subsequent deposition is the major source of elevated Hg observed in the mushrooms, litter and surface layer of soils in the outskirts of Gongga Mountain maritime glacier that has a peak of 7556m above sea level.
Sigma-Aldrich
Mercury, ACS reagent, 99.9995% trace metals basis