Merck
  • Ghrelin acts as an interface between physiological state and phasic dopamine signaling.
Ghrelin acts as an interface between physiological state and phasic dopamine signaling.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-04-04)
Jackson J Cone, James E McCutcheon, Mitchell F Roitman
ABSTRACT
Brief, high-concentration (phasic) spikes in nucleus accumbens dopamine critically participate in aspects of food reward. Although physiological state (e.g., hunger, satiety) and associated hormones are known to affect dopamine tone in general, whether they modulate food-evoked, phasic dopamine specifically is unknown. Here, we used fast-scan cyclic voltammetry in awake, behaving rats to record dopamine spikes evoked by delivery of sugar pellets while pharmacologically manipulating central receptors for the gut "hunger" hormone ghrelin. Lateral ventricular (LV) ghrelin increased, while LV ghrelin receptor antagonism suppressed the magnitude of dopamine spikes evoked by food. Ghrelin was effective when infused directly into the lateral hypothalamus (LH), but not the ventral tegmental area (VTA). LH infusions were made in close proximity to orexin neurons, which are regulated by ghrelin and project to the VTA. Thus, we also investigated and found potentiation of food-evoked dopamine spikes by intra-VTA orexin-A. Importantly, intra-VTA blockade of orexin receptors attenuated food intake induced by LV ghrelin, thus establishing a behaviorally relevant connection between central ghrelin and VTA orexin. Further analysis revealed that food restriction increased the magnitude of dopamine spikes evoked by food independent of any pharmacological manipulations. The results support the regulation of food-evoked dopamine spikes by physiological state with endogenous fluctuations in ghrelin as a key contributor. Our data highlight a novel mechanism by which signals relating physiological state could influence food reinforcement and food-directed behavior.
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Dopamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Dopamine hydrochloride
Supelco
Dopamine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Urea, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Supelco
Dopamine hydrochloride solution, 1.0 mg/mL in methanol with 5% 1 M HCl (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Urea, analytical standard