Merck
  • [Investigation of optimum concentrations of betaine for improving the resolution of sequencing G-C rich DNA with trinucleotide repeats].
[Investigation of optimum concentrations of betaine for improving the resolution of sequencing G-C rich DNA with trinucleotide repeats].
Zhonghua yi xue yi chuan xue za zhi = Zhonghua yixue yichuanxue zazhi = Chinese journal of medical genetics (2014-04-09)
Dan Wang, Hao Cai, Long Yu
ABSTRACT
To develop an optimal sequencing system which can improve the resolution of sequencing G-C rich DNA with abundant trinucleotide repeats by applying concentration gradients of betaine to the Sanger sequencing system. Concentration gradients of betaine were introduced into the sequencing system by taking the 5' terminal of Nogo-B cDNA (Am-Nogo-B) (G-C%=72%, without trinucleotide repeats) and 5' terminal of Huntingtin cDNA (Am-HTT) (G-C%=74%, with abundant CAG and CCG repeats) the results of sequencing were compared. The optimum concentration of betaine for sequencing Am-Nogo-B has differed from that for Am-HTT. Result of sequencing Am-Nogo-B has achieved the best quality when the concentration of betaine was at 0.8-1.2 mol/L, whereas the result of sequencing Am-HTT obtained the best quality when the concentration of betaine was at 1.6 -2.4 mol/L. The results were reproducible. G-C rich DNA with similar G-C% required different concentrations of betaine in the sequencing system due to base pair compositions. The sequencing system developed for improving the resolution of sequencing of G-C rich DNA with abundant trinucleotide repeats can be used as a reference for similar studies.
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
Sigma-Aldrich
Betaine hydrochloride, ≥99%
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent