Merck
  • Novel chitosan coated magnetic nanocarriers for the targeted Diclofenac delivery.
Novel chitosan coated magnetic nanocarriers for the targeted Diclofenac delivery.
Journal of nanoscience and nanotechnology (2014-04-17)
Mariela Agotegaray, Santiago Palma, Verónica Lassalle
ABSTRACT
New magnetic devices consisting of magnetite functionalized with oleic acid and chitosan have been synthesized and employed to the loading of Diclofenac as potential tool for treatment of targeted inflammatory diseases. Magnetic loaded and un-loaded nanoparticles have been thoroughly characterized by infrared spectroscopy, transmission electron microscopy, determination of hydrodynamic diameter by Dynamic light scattering and zeta potential measurements at different pH conditions. A study of the release of Diclofenac has been performed in vitro and available mathematical models have been used to determine the release kinetic. Both properties and release data reveal that this nanomagnetic platform would be suitable for in vivo assays.
Sigma-Aldrich
Chitosan, high molecular weight
Diclofenac potassium, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Diclofenac sodium salt
Supelco
Diclofenac sodium salt, analytical standard
Supelco
Diclofenac sodium salt, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chitosan, from shrimp shells, practical grade
Sigma-Aldrich
Chitosan, low molecular weight
Sigma-Aldrich
Chitosan, from shrimp shells, ≥75% (deacetylated)
Sigma-Aldrich
Chitosan, medium molecular weight
Diclofenac for system suitability, European Pharmacopoeia (EP) Reference Standard
Diclofenac sodium, European Pharmacopoeia (EP) Reference Standard
USP
Diclofenac sodium, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Chitosan from shrimp shells, low-viscous