Merck
  • Nanostructured hybrid of immiscible gold and silicon and its effect on proliferation and adhesion of fibroblasts and osteoblasts.
Nanostructured hybrid of immiscible gold and silicon and its effect on proliferation and adhesion of fibroblasts and osteoblasts.
Journal of biomedical nanotechnology (2014-04-23)
Priyatha Premnath, Bo Tan, Krishnan Venkatakrishnan
ABSTRACT
Hybrid biomaterials are a combination of two or more different materials that work synergistically to produce superior properties. Nano structuring of such hybrid materials has also posed complications. In this study, we present, for the first time a nanofibrous hybrid of gold and silicon fabricated by femtosecond laser synthesis for tissue engineering applications. The formation of a completely new phase, Au3Si (212) is reported. The formation mechanism is explained by vapor condensation. Particle sizes of 2-10 nm and 37-49 nm for gold and gold concentrations of 35-78% are achieved. The effect of this hybrid on cell growth was assessed using fibroblasts and osteoblasts. There was a significant decrease in both osteoblast and fibroblast proliferation with the increase of gold in the hybrid nanostructure. This novel hybrid nanofibrous matrix provides a method to effectively control the proliferation and adhesion of cells. Femtosecond laser synthesis presents a new standard by which not only a single element biomaterial but also multiple immiscible element hybrid biomaterials can be fabricated. This technique provides a paradigm shift in the fabrication of novel nanostructured immiscible hybrid biomaterials.
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer, <111>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.3 mm
Sigma-Aldrich
Gold coated glass cover slip, L × W × thickness 22 mm × 22 mm × 130-170 μm, square, layer thickness 100 Å, 99.999% (Au)
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Silicon, rod, 40mm, diameter 20mm, single crystal, -100, 99.999%
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, nanopowder, <100 nm particle size (TEM), ≥98% trace metals basis
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.5 mm
Silicon, rod, 100mm, diameter 12.7mm, single crystal - random orientation, 100%
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), contains phosphorus as dopant, <111>, N-type, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Gold, foil, thickness 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold coated glass cover slip, 99.999% (Au), diam. × thickness 15 mm × 130-170 μm, layer thickness 100 Å
Sigma-Aldrich
Gold coated glass slide, layer thickness 100 Å, 99.999% (Au)
Gold, insulated wire, 1m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyester insulation, 99.99%
Sigma-Aldrich
Gold coated glass slide, layer thickness 1000 Å, 99.999% (Au)
Sigma-Aldrich
Silicon, pieces, 99.95% trace metals basis
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.3 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
Silicon, powder, −325 mesh, 99% trace metals basis
Sigma-Aldrich
Gold, powder, <850 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.025 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold, nanopowder, <100 nm particle size, 99.9% trace metals basis