Merck
  • The use of local agents: bone wax, gelatin, collagen, oxidized cellulose.
The use of local agents: bone wax, gelatin, collagen, oxidized cellulose.
European spine journal : official publication of the European Spine Society, the European Spinal Deformity Society, and the European Section of the Cervical Spine Research Society (2004-06-29)
Claudio Schonauer, Enrico Tessitore, Giuseppe Barbagallo, Vincenzo Albanese, Aldo Moraci
ABSTRACT
The use of local agents to achieve hemostasis is an old and complex subject in surgery. Their use is almost mandatory in spinal surgery. The development of new materials in chemical hemostasis is a continuous process that may potentially lead the surgeon to confusion. Moreover, the more commonly used materials have not changed in about 50 years. Using chemical agents to tamponade a hemorrhage is not free of risks. Complications are around the corner and can be due either to mechanical compression or to phlogistic effects secondary to the material used. This paper reviews about 20 animal and clinical published studies with regard to the chemical properties, mechanisms of action, use and complications of local agents.
Sigma-Aldrich
Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
Gelatin from bovine skin, gel strength ~225 g Bloom, Type B
Sigma-Aldrich
Gelatin from bovine skin, Type B
Sigma-Aldrich
Gelatin, tested according to Ph. Eur.
Sigma-Aldrich
Gelatin from porcine skin, gel strength 80-120 g Bloom, Type A
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Sigma-Aldrich
Gelatin from porcine skin, gel strength ~175 g Bloom, Type A
Sigma-Aldrich
Prionex® Highly purified Type A, aqueous solution
Sigma-Aldrich
Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Gelatin from cold water fish skin, solid
Millipore
Gelatin from porcine skin, suitable for microbiology, high gel strength
Millipore
Gelatin from porcine skin, suitable for microbiology, low gel strength
Millipore
Gelatin from porcine skin, medium gel strength, suitable for microbiology
Millipore
Gelatin from porcine skin, suitable for microbiology, ultrahigh gel strength
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture