Merck
Fragrance material review on farnesol.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2008-07-22)
A Lapczynski, S P Bhatia, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of farnesol when used as a fragrance ingredient is presented.
Sigma-Aldrich
Farnesol, mixture of isomers, ≥95%, stabilized, FG
Sigma-Aldrich
Farnesol, 95%
Supelco
Farnesol, stabilized, analytical standard, mixture of isomers
Supelco
Farnesol, mixture of isomers, analytical standard