Merck
Dapsone-induced photosensitivity: a rare clinical presentation.
Photodermatology, photoimmunology & photomedicine (2008-09-25)
Bikash Ranjan Kar
ABSTRACT
Dapsone is an efficient anti-inflammatory and antimycobacterial agent. It is one of the main constituents of multidrug therapy (MDT). It acts by interference with folate metabolism. Dapsone-induced photosensitivity is a rare, non-dose-related adverse effect of the sulfone and can occur in patients with inflammatory skin disorders treated with dapsone. So far, only 12 cases seem to have been reported in the literature. We report a case of dapsone-induced photosensitivity in an Indian patient with leprosy.
Product Description
Supelco
Dapsone, VETRANAL®, analytical standard
Dapsone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
4-Aminophenyl sulfone, 97%