Merck
  • Modified citrus pectin anti-metastatic properties: one bullet, multiple targets.
Modified citrus pectin anti-metastatic properties: one bullet, multiple targets.
Carbohydrate research (2008-12-09)
Vladislav V Glinsky, Avraham Raz
ABSTRACT
In this minireview, we examine the ability of modified citrus pectin (MCP), a complex water soluble indigestible polysaccharide obtained from the peel and pulp of citrus fruits and modified by means of high pH and temperature treatment, to affect numerous rate-limiting steps in cancer metastasis. The anti-adhesive properties of MCP as well as its potential for increasing apoptotic responses of tumor cells to chemotherapy by inhibiting galectin-3 anti-apoptotic function are discussed in the light of a potential use of this carbohydrate-based substance in the treatment of multiple human malignancies.
Sigma-Aldrich
Pectin from citrus peel, Galacturonic acid ≥74.0 % (dried basis)
Supelco
Pectin, for use with Total Dietary Fiber Control Kit, TDF-C10